Lutetium trichloride, ultra dry ≥99.98% (REO, rare earth oxide basis)
Dodavatel: Thermo Scientific
Ampuled under argon
35802.03
35802.06
35802.03EA
5120
CZK
35802.03
35802.06
Lutetium trichloride, ultra dry ≥99.98% (REO, rare earth oxide basis)
Lutetium trichloride
Purity: 99.98%
CAS No.: 10099-66-8
Molecular Formula: LuCl3
Formula Weight: 281.33
Physical Form: Powder, Ampouled under argon
Density: 3.98
Melting Point: 905° subl.
Merck Reference: 14,5615
MDL No.: MFCD00011099
Vzorec:
Cl₃Lu MW: 281,33 g/mol Bod tání: 905 °C (subl.) Hustota: 3,98 Storage Temperature: Ambient |
MDL Number:
MFCD00011099 CAS číslo: 10099-66-8 EINECS: 233-240-1 Merck Index: 14,5615 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...