Tisk…

Lanthanum(III) chloride, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Lanthanum chloride, Lanthanum trichloride

M-Clarity™ quality level = MQ100

449830-25G 449830-5G
SIAL449830-25GEA 10630 CZK
SIAL449830-25G SIAL449830-5G
Lanthanum(III) chloride, Sigma-Aldrich®
Lanthanum(III) chloride
Vzorec: Cl₃La
MW: 245.26
Bod tání: 857
Hustota: 3.8773 g/cm³ (–0.15 °C)
MDL Number: MFCD00011068
CAS číslo: 10099-58-8
EINECS: 233-237-5
Merck Index: 13,05379

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR