Lanthanum(III) chloride 99.9%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Lanthanum chloride, Lanthanum trichloride
315230100
315230500
315232500
ACRO315230100EA
3420
CZK
ACRO315230100
ACRO315230500
ACRO315232500
Lanthanum(III) chloride 99.9%
Lanthanum(III) chloride
Vzorec:
Cl₃La Bod varu: 1000 °C Bod tání: 860 °C Hustota: 3,8773 g/cm³ (-0.15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011068 CAS číslo: 10099-58-8 EINECS: 233-237-5 UN: 1759 ADR: 8,III Merck Index: 13,05379 |
Výsledky specifikačního testu
Appearance | White to off-white Powder |
Total conc. of imp.as REO | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...