Tisk…

Holmium(III) chloride, anhydrous ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Holmium trichloride

18681.06 18681.14
18681.06EA 5940 CZK
18681.06 18681.14
Holmium(III) chloride, anhydrous ≥99.9% (metals basis)
Holmium(III) chloride

Purity: 99.9% CAS No.: 10138-62-2 Molecular Formula: HoCl3 Formula Weight: 271.29 Physical Form: -20 Mesh Density: 3.7 Melting Point: 718° Merck Reference: 14,4727 MDL No.: MFCD00011051

Vzorec: Cl₃Ho
Bod varu: 1500 °C (1013 hPa)
Bod tání: 718 °C
Hustota: 3,7
Storage Temperature: Ambient
MDL Number: MFCD00011051
CAS číslo: 10138-62-2
EINECS: 233-387-1
Merck Index: 14,4727

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR