Tisk…

Europium(III) chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

238066-10G 238066-1G
SIAL238066-10GEA 16690 CZK
SIAL238066-10G SIAL238066-1G
Europium(III) chloride, Sigma-Aldrich®
Europium(III) chloride
Vzorec: Cl₃Eu
MW: 258,32 g/mol
Bod tání: 626…638 °C
Hustota: 4,89 g/cm³ (20 °C)
MDL Number: MFCD00010994
CAS číslo: 10025-76-0
EINECS: 233-040-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR