Cerium trichloride, anhydrous ≥99.5% (REO, rare earth oxide basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Cerous chloride, Cerium(III) chloride, Cerium (III) chloride
Purity: 99.5%
CAS No.: 7790-86-5
Molecular Formula: CeCl3
Formula Weight: 246.48
Physical Form: Powder
Density: 3.92
Appearance: White to pale grey
Odour: Odourless
Melting Point: 848°
Boiling Point: 1727°
Solubility: Soluble in water, alcohol, acetone, acids
Merck Reference: 14,1997
MDL No.: MFCD00010929
Incandescent gas mantles, spectrography, polymerization of olefins
Vzorec:
CeCl₃ MW: 246.47 Bod varu: 1727 °C Bod tání: 848 Hustota: 3.92 Storage Temperature: Ambient |
MDL Number:
MFCD00010929 CAS číslo: 7790-86-5 EINECS: 232-227-8 Merck Index: 14,1997 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...