Thulium(III) bromide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Dodavatel: Thermo Scientific
|
Varování
|
Ampuled under argon
39186.03
39186.06
39186.03EA
4950
CZK
39186.03
39186.06
Thulium(III) bromide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Thulium(III) bromide
Purity: 99.99%
CAS No.: 14456-51-0
Molecular Formula: TmBr3
Formula Weight: 408.66
Physical Form: Powder, Ampouled under argon
Sensitivity: Hygroscopic
Boiling Point: 1440°
MDL No.: MFCD00050138
Vzorec:
Br₃Tm MW: 408,65 g/mol Bod varu: 1440 °C Bod tání: 952 °C Storage Temperature: Ambient |
MDL Number:
MFCD00050138 CAS číslo: 14456-51-0 EINECS: 238-444-4 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...