Lanthanum(III) bromide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Ampuled under argon
44396.09
44396.18
44396.09EA
1700
CZK
44396.09
44396.18
Lanthanum(III) bromide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Lanthanum(III) bromide
Purity: 99.9%
CAS No.: 13536-79-3
Molecular Formula: LaBr3
Formula Weight: 378.62
Physical Form: Lumps, Ampouled under argon
Sensitivity: Hygroscopic
Appearance: White
MDL No.: MFCD00016161
Vzorec:
Br₃La MW: 378.62 Bod tání: 789 Hustota: 5.06 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016161 CAS číslo: 13536-79-3 EINECS: 236-896-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...