Tisk…

Cerium tribromide ≥99.998% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Cerous bromide, Cerium(III) bromide hydrate, Cerium (III) bromide

44432.09 44432.18
44432.09EA 2780 CZK
44432.09 44432.18
Cerium tribromide ≥99.998% (metals basis)
Cerium tribromide

Purity: 99.998% CAS No.: 14457-87-5 Molecular Formula: CeBr3.xH2O Formula Weight: 379.83(anhy) Physical Form: Crystalline Appearance: Pale orange Odour: Odourless Merck Reference: 14,1995 MDL No.: MFCD00150206

Vzorec: CeBr₃
MW: 379.83
Bod tání: 732
Storage Temperature: Ambient
MDL Number: MFCD00016004
CAS číslo: 14457-87-5
EINECS: 238-447-0
Merck Index: 14,1995

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR