Tisk…

Cerium tribromide, ultra dry ≥99.9% (REO, rare earth oxide basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Cerous bromide, Cerium(III) bromide hydrate, Cerium (III) bromide

Ampuled under argon

40373.03 40373.06 40373.14
40373.03EA 3350 CZK
40373.03 40373.06 40373.14
Cerium tribromide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Cerium tribromide

Purity: 99.9% CAS No.: 14457-87-5 Molecular Formula: CeBr3 Formula Weight: 379.83 Physical Form: Powder, Ampouled under argon Density: 5.18 Appearance: White to pale grey Odour: Odourless Melting Point: 733° Boiling Point: 1560° Merck Reference: 14,1995 MDL No.: MFCD00016004

Vzorec: CeBr₃
MW: 379.83
Bod varu: 1560 °C
Bod tání: 733 °C
Hustota: 5.18
Storage Temperature: Ambient
MDL Number: MFCD00016004
CAS číslo: 14457-87-5
EINECS: 238-447-0
Merck Index: 14,1995

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR