Tisk…

6-Methylindole ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 6-Methyl-1H-indole

L17509.03 L17509.MD
L17509.03EA 2970 CZK
L17509.03 L17509.MD
6-Methylindole ≥98%
6-Methylindole

Purity: 98% CAS No.: 3420-02-8 Molecular Formula: C9H9N Formula Weight: 131.18 Flash Point: 110°(230°F) Refractive Index: 1.6071 Melting Point: 29-32° Boiling Point: 112°/5mm Beilstein Reference: 109708 MDL No.: MFCD00005682

Vzorec: C₉H₉N
MW: 131,18 g/mol
Bod varu: 112 °C (5 mmHg)
Bod tání: 29…32 °C
Hustota: 1,059
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005682
CAS číslo: 3420-02-8
UN: 3334
ADR: 9,

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR