Tisk…

1-Methylindole ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: N-Methylindole, 1-Methyl-1H-indole

A12605.09 A12605.18
A12605.09EA 2320 CZK
A12605.09 A12605.18
1-Methylindole ≥98%
1-Methylindole

Purity: 98% CAS No.: 603-76-9 Molecular Formula: C9H9N Formula Weight: 131.18 Flash Point: 110°(230°F) Refractive Index: 1.6065 Odour: Stench Boiling Point: 237-239° Beilstein Reference: 111026 MDL No.: MFCD00005800

Vzorec: C₉H₉N
Bod varu: 237…239 °C
Bod tání: 94…95 °C
Hustota: 1,048
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005800
CAS číslo: 603-76-9
EINECS: 210-057-5
UN: 3334
ADR: 9,

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR