5-Nitroindole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
5-Nitro-1H-indole
M-Clarity™ quality level = MQ100
N17602-25G
N17602-5G
SIALN17602-25GEA
4590
CZK
SIALN17602-25G
SIALN17602-5G
5-Nitroindole, Sigma-Aldrich®
5-Nitroindole
Vzorec:
C₈H₆N₂O₂ MW: 162,15 g/mol Bod tání: 140 °C |
MDL Number:
MFCD00005673 CAS číslo: 6146-52-7 EINECS: 228-153-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...