Tisk…

5-Nitroindole ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 5-Nitro-1H-indole

A10608.03 A10608.06 A10608.14
A10608.03EA 900 CZK
A10608.03 A10608.06 A10608.14
5-Nitroindole ≥99%
5-Nitroindole

Purity: 99% CAS No.: 6146-52-7 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Melting Point: 139-143° Beilstein Reference: 383777

Vzorec: C₈H₆N₂O₂
MW: 162,15 g/mol
Bod tání: 139…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00005673
CAS číslo: 6146-52-7
EINECS: 228-153-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR