7-Fluoroindole ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
L17621.03
L17621.MD
L17621.03EA
1840
CZK
L17621.03
L17621.MD
7-Fluoroindole ≥97%
7-Fluoroindole
Purity: 97%
CAS No.: 387-44-0
Molecular Formula: C8H6FN
Formula Weight: 135.14
Melting Point: 61-63°
Beilstein Reference: 114740
Vzorec:
C₈H₆FN MW: 135,14 g/mol Bod tání: 61…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074502 CAS číslo: 387-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...