Indole-5-carboxylic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1H-Indole-5-carboxylic acid
M-Clarity™ quality level = MQ100
I5400-1G
I5400-5G
SIALI5400-1GEA
0
CZK
SIALI5400-1G
SIALI5400-5G
Indole-5-carboxylic acid, Sigma-Aldrich®
Indole-5-carboxylic acid
Vzorec:
C₉H₇NO₂ MW: 161,16 g/mol Bod tání: 211…213 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005678 CAS číslo: 1670-81-1 EINECS: 216-799-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...