Indole-3-carboxylic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1H-Indole-3-carboxylic acid
A12634.03
A12634.06
A12634.14
A12634.03EA
1240
CZK
A12634.03
A12634.06
A12634.14
Indole-3-carboxylic acid ≥98%
Indole-3-carboxylic acid
Purity: 98%
CAS No.: 771-50-6
Molecular Formula: C9H7NO2
Formula Weight: 161.16
Melting Point: ca 239° dec.
Beilstein Reference: 129435
Vzorec:
C₉H₇NO₂ MW: 161,16 g/mol Bod tání: ∼ 239 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00005624 CAS číslo: 771-50-6 EINECS: 212-231-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...