Indole-2-carboxylic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1H-Indole-2-carboxylic acid
M-Clarity™ quality level = MQ100
I5109-10G
I5109-50G
I5109-5G
SIALI5109-10GEA
2120
CZK
SIALI5109-10G
SIALI5109-50G
SIALI5109-5G
Indole-2-carboxylic acid, Sigma-Aldrich®
Indole-2-carboxylic acid
Vzorec:
C₉H₇NO₂ MW: 161,16 g/mol Bod tání: 203…206 °C |
MDL Number:
MFCD00005611 CAS číslo: 1477-50-5 EINECS: 216-030-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...