Indole-5-carbonitrile, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1H-Indole-5-carbonitrile, 5-Cyanoindole, 5-Cyano-1H-indole
M-Clarity™ quality level = MQ100
C92006-10G
C92006-5G
SIALC92006-10GEA
12240
CZK
SIALC92006-10G
SIALC92006-5G
Indole-5-carbonitrile, Sigma-Aldrich®
Indole-5-carbonitrile
Vzorec:
C₉H₆N₂ MW: 142,16 g/mol Bod tání: 106…108 °C |
MDL Number:
MFCD00005669 CAS číslo: 15861-24-2 EINECS: 239-986-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...