Tisk…

7-Chloroindole ≥98%

Dodavatel: Thermo Scientific
Varování

L17637.03 L17637.MD
L17637.03EA 4120 CZK
L17637.03 L17637.MD
7-Chloroindole ≥98%
7-Chloroindole

Purity: 98% CAS No.: 53924-05-3 Molecular Formula: C8H6ClN Formula Weight: 151.60 Melting Point: 57-58° Beilstein Reference: 114743

Vzorec: C₈H₆ClN
MW: 151,6 g/mol
Bod tání: 57…58 °C
Storage Temperature: Ambient
MDL Number: MFCD01321301
CAS číslo: 53924-05-3
EINECS: 258-865-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR