5-Chloroindole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
5-Chloro-1H-indole
M-Clarity™ quality level = MQ100
C47604-1G
C47604-5G
SIALC47604-1GEA
2530
CZK
SIALC47604-1G
SIALC47604-5G
5-Chloroindole, Sigma-Aldrich®
5-Chloroindole
Vzorec:
C₈H₆ClN MW: 151,6 g/mol Bod tání: 71…72 °C |
MDL Number:
MFCD00005672 CAS číslo: 17422-32-1 EINECS: 241-448-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...