5-Bromoindole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
5-Bromo-1H-indole
M-Clarity™ quality level = MQ100
B68607-25G
B68607-5G
SIALB68607-25GEA
4910
CZK
SIALB68607-25G
SIALB68607-5G
5-Bromoindole, Sigma-Aldrich®
5-Bromoindole
Vzorec:
C₈H₆BrN Bod tání: 90…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005670 CAS číslo: 10075-50-0 EINECS: 233-208-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...