Tisk…

Inden ≥90% stabilizovaný

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A12192.AD A12192.AK A12192.AU
A12192.ADEA 1240 CZK
A12192.AD A12192.AK A12192.AU
Inden ≥90% stabilizovaný
Inden

Stabilized with 0.01% 4-tert-butylcatechol


Purity: 90+% CAS No.: 95-13-6 Molecular Formula: C9H8 Formula Weight: 116.16 Storage Temperature: 2°C to 8°C Flash Point: 58°(136°F) Refractive Index: 1.5700 Odour: Aromatic Melting Point: ca -2° Boiling Point: 181-182° Beilstein Reference: 635873 Merck Reference: 14,4939 UN No.: 3295 MDL No.: MFCD00003777

Vzorec: C₉H₈
Bod varu: 181…182 °C
Bod tání: ∼–2 °C
Hustota: 0,996
Bod vzplanutí: 58 °C (136 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00003777
CAS číslo: 95-13-6
EINECS: 202-393-6
UN: 3295
ADR: 3,III
Merck Index: 14,4939

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR