5-Fluoro-1-indanone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
5-Fluoro-2,3-dihydro-1H-inden-1-one
M-Clarity™ quality level = MQ100
185663-1G
185663-5G
SIAL185663-1GEA
3080
CZK
SIAL185663-1G
SIAL185663-5G
5-Fluoro-1-indanone, Sigma-Aldrich®
5-Fluoro-1-indanone
Vzorec:
C₉H₇FO MW: 150,15 g/mol Bod varu: 113…114 °C (10 mmHg) Bod tání: 35…37 °C Hustota: 1,35 g/cm³ (20 °C) |
MDL Number:
MFCD00041031 CAS číslo: 700-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...