Tisk…

1,3-Indandione, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,3-Indanedione, Indan-1,3-dione, 1H-Indene-1,3(2H)-dione

M-Clarity™ quality level = MQ200

I2002-5G-A I2002-25G-A
SIALI2002-5G-AEA 2150 CZK
SIALI2002-5G-A SIALI2002-25G-A
1,3-Indandione, Sigma-Aldrich®
1,3-Indandione
Vzorec: C₉H₆O₂
MW: 146,15 g/mol
Bod varu: 283 °C (1013 hPa)
Bod tání: 129…132 °C
Hustota: 1,37 g/cm³ (21 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003779
CAS číslo: 606-23-5
EINECS: 210-109-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR