Tisk…

5-Methoxy-1-indanone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

183539-10G 183539-5G
SIAL183539-10GEA 9520 CZK
SIAL183539-10G SIAL183539-5G
5-Methoxy-1-indanone, Sigma-Aldrich®
5-Methoxy-1-indanone
Vzorec: C₁₀H₁₀O₂
MW: 162,19 g/mol
Bod varu: 145…150 °C (0,2 mmHg)
Bod tání: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00003789
CAS číslo: 5111-70-6
EINECS: 225-838-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR