Tisk…

2-Indanone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: beta-Hydrindone, 2-Oxoindane, 1H-Inden-2(3H)-one

M-Clarity™ quality level = MQ100

146692-10G 146692-50G
SIAL146692-10GEA 4610 CZK
SIAL146692-10G SIAL146692-50G
2-Indanone, Sigma-Aldrich®
2-Indanone
Vzorec: C₉H₈O
MW: 132,16 g/mol
Bod varu: 220…225 °C (1013 hPa)
Bod tání: 54…56 °C
Hustota: 1,0712 g/cm³ (67 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003792
CAS číslo: 615-13-4
EINECS: 210-410-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR