Tisk…

1,2-Dimethylimidazole ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1,2-Dimethyl-1H-imidazole

B23544.14 B23544.22 B23544.36
B23544.14EA 704 CZK
B23544.14 B23544.22 B23544.36
1,2-Dimethylimidazole ≥98%
1,2-Dimethylimidazole

Purity: 98% CAS No.: 1739-84-0 Molecular Formula: C5H8N2 Formula Weight: 96.13 Flash Point: 92°(197°F) Melting Point: 35-38° Boiling Point: 203-205° Beilstein Reference: 108457 MDL No.: MFCD00005294

Vzorec: C₅H₈N₂
MW: 96,13 g/mol
Bod varu: 203…205 °C
Bod tání: 35…38 °C
Hustota: 1,084
Bod vzplanutí: 92 °C (197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005294
CAS číslo: 1739-84-0
EINECS: 217-101-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR