Tisk…

2-Nitroimidazole ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Azomycin, 2-Nitro-1H-imidazole

L13546.MC L13546.ME
L13546.MCEA 1900 CZK
L13546.MC L13546.ME
2-Nitroimidazole ≥98%
2-Nitroimidazole

Purity: 98% CAS No.: 527-73-1 Molecular Formula: C3H3N3O2 Formula Weight: 113.08 Melting Point: ca 287° dec. Beilstein Reference: 116444 Merck Reference: 14,921 UN No.: 2811 MDL No.: MFCD00005185

Vzorec: C₃H₃N₃O₂
MW: 113,08 g/mol
Bod tání: ∼ 287 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00005185
CAS číslo: 527-73-1
EINECS: 208-425-5
UN: 2811
ADR: 6.1,III
Merck Index: 14,921

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR