Tisk…

Fenylhydrazin 97%

Dodavatel: Thermo Fisher Scientific

296685000 296681000 296680025
ACRO296685000EA 3070 CZK
ACRO296685000 ACRO296681000 ACRO296680025
Fenylhydrazin 97%
Fenylhydrazin

Objednat

Výsledky specifikačního testu

Appearance Clear yellow to light orange Liquid
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR