Tisk…

Oxid jodičný ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: di-Iodine pentaoxide, Iodopentoxide, di-Iodine pentoxide

87682.14 87682.22
87682.14EA 2600 CZK
87682.14 87682.22
Oxid jodičný ≥98%
Oxid jodičný

Purity: 98% CAS No.: 12029-98-0 Molecular Formula: I2O5 Formula Weight: 333.81 Physical Form: Crystalline Density: 5.08 Appearance: White to cream pH: 1.1 Melting Point: 275-350° dec. Solubility: Soluble in water and HNO3. Insoluble in absolute alcohol, chloroform, ether, and CS2 Merck Reference: 14,5019 UN No.: 3085 MDL No.: MFCD00011356


An oxidizing agent to remove CO from gas

Vzorec: I₂O₅
Hustota: 5,08
Storage Temperature: Ambient
MDL Number: MFCD00011356
CAS číslo: 12029-98-0
EINECS: 234-740-2
UN: 3085
ADR: 5.1,II
Merck Index: 14,5019

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR