Diphenyl selenide ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A18364.03
A18364.06
A18364.14
A18364.03EA
1060
CZK
A18364.03
A18364.06
A18364.14
Diphenyl selenide ≥98%
Diphenyl selenide
Purity: 98%
CAS No.: 1132-39-4
Molecular Formula: (C6H5)2Se
Formula Weight: 233.17
Odour: Unpleasant
Melting Point: 3°
Boiling Point: 165-167°/12mm
UN No.: 3440
MDL No.: MFCD00014067
Vzorec:
C₁₂H₁₀Se Bod varu: 165…167 °C (12 mmHg) Bod tání: 3 °C Hustota: 1,351 Storage Temperature: Ambient |
MDL Number:
MFCD00014067 CAS číslo: 1132-39-4 EINECS: 214-474-2 UN: 3440 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...