Tisk…

Diphenyl ditelluride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

384127-1G 384127-5G
SIAL384127-1GEA 1960 CZK
SIAL384127-1G SIAL384127-5G
Diphenyl ditelluride, Sigma-Aldrich®
Diphenyl ditelluride
Vzorec: C₁₂H₁₀Te₂
MW: 409.41
Bod tání: 67
MDL Number: MFCD00192106
CAS číslo: 32294-60-3
EINECS: 250-982-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR