Diphenyl diselenide, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
180629-100G
180629-25G
180629-5G
SIAL180629-100GEA
19460
CZK
SIAL180629-100G
SIAL180629-25G
SIAL180629-5G
Diphenyl diselenide, Sigma-Aldrich®
Diphenyl diselenide
Vzorec:
C₁₂H₁₀Se₂ Bod varu: 377 °C (1013 hPa) Bod tání: 58…61 °C Hustota: 1,32 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003001 CAS číslo: 1666-13-3 EINECS: 216-780-2 UN: 3283 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...