Dibenzyl disulphide ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Benzyl disulphide
L11850.14
L11850.22
L11850.14EA
906
CZK
L11850.14
L11850.22
Dibenzyl disulphide ≥98%
Dibenzyl disulphide
Purity: 98+%
CAS No.: 150-60-7
Molecular Formula: C6H5CH2S-SCH2C6H5
Formula Weight: 246.40
Flash Point: 150°(302°F)
Odour: Stench
Boiling Point: 210-216°/18mm
Beilstein Reference: 1110443
Merck Reference: 14,3013
MDL No.: MFCD00004783
Vzorec:
C₁₄H₁₄S₂ MW: 246,4 g/mol Bod varu: 210…216 °C (18 mmHg) Bod tání: 67…72 °C Hustota: 1,300 Bod vzplanutí: 150 °C (302 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004783 CAS číslo: 150-60-7 EINECS: 205-764-0 Merck Index: 14,3013 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...