Tisk…

Dioctadecyl disulphide ≥98%

Dodavatel: Thermo Scientific

B21978.03 B21978.06 B21978.14
B21978.03EA 1140 CZK
B21978.03 B21978.06 B21978.14
Dioctadecyl disulphide ≥98%
Dioctadecyl disulphide

Purity: 98% CAS No.: 2500-88-1 Molecular Formula: CH3(CH2)17S-S(CH2)17CH3 Formula Weight: 571.11 Odour: Unpleasant Melting Point: 60-64° Beilstein Reference: 1715376 MDL No.: MFCD00026687

Vzorec: C₃₆H₇₄S₂
MW: 571,12 g/mol
Bod tání: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00026687
CAS číslo: 2500-88-1
EINECS: 219-702-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR