Dimethyldisulfid, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Dimethyl disulphide, Methyl disulphide
M-Clarity™ quality level = MQ300
W353604-10KG-K
W353604-1KG-K
W353604-5KG-K
SIALW353604-10KG-KEA
14440
CZK
SIALW353604-10KG-K
SIALW353604-1KG-K
SIALW353604-5KG-K
Dimethyldisulfid, Sigma-Aldrich®
Dimethyldisulfid
Vzorec:
C₂H₆S₂ MW: 94,2 g/mol Bod varu: 110 °C (1013 hPa) Bod tání: –98 °C Hustota: 1,06 g/cm³ (20 °C) Bod vzplanutí: 16 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008561 CAS číslo: 624-92-0 EINECS: 210-871-0 UN: 2381 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...