Diethyl disulphide, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Ethyldisulphide
M-Clarity™ quality level = MQ100
E26223-100G
E26223-25G
SIALE26223-100GEA
4440
CZK
SIALE26223-100G
SIALE26223-25G
Diethyl disulphide, Sigma-Aldrich®
Diethyl disulphide
Vzorec:
C₄H₁₀S₂ Bod varu: 152 °C (1013 hPa) Hustota: 0,984 g/cm³ (20 °C) |
MDL Number:
MFCD00009266 CAS číslo: 110-81-6 EINECS: 203-805-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...