Diethyl disulphide ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Ethyldisulphide
L10065.14
L10065.22
L10065.14EA
1800
CZK
L10065.14
L10065.22
Diethyl disulphide ≥99%
Diethyl disulphide
Purity: 99%
CAS No.: 110-81-6
Molecular Formula: C4H10S2
Formula Weight: 122.25
Flash Point: 40°(104°F)
Refractive Index: 1.5060
Odour: Stench
Melting Point: -102°
Boiling Point: 152-154°
Beilstein Reference: 1098273
UN No.: 1993
MDL No.: MFCD00009266
Vzorec:
C₄H₁₀S₂ MW: 122,26 g/mol Bod varu: 152…154 °C Bod tání: –102 °C Hustota: 0,993 Bod vzplanutí: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009266 CAS číslo: 110-81-6 EINECS: 203-805-7 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...