Tisk…

Diallyl disulphide, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ300

W202800-100G-K W202800-1KG-K W202800-5KG-K
SIALW202800-100G-KEA 3780 CZK
SIALW202800-100G-K SIALW202800-1KG-K SIALW202800-5KG-K
Diallyl disulphide, Sigma-Aldrich®
Diallyl disulphide
Vzorec: C₆H₁₀S₂
Bod varu: 198 °C (1013 hPa)
Hustota: 1,0237 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00008656
CAS číslo: 2179-57-9
EINECS: 218-548-6
UN: 2810
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR