Sulfamát amonný ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Ammonium sulphamidate, Ammonium amidosulphonate
A17696.30
A17696.0B
A17696.30EA
1070
CZK
A17696.30
A17696.0B
Sulfamát amonný ≥98%
Ammonium sulphamate
Purity: 98+%
CAS No.: 7773-06-0
Molecular Formula: H6N2O3S
Formula Weight: 114.12
Odour: Weak
pH: 5.2
Melting Point: 131-135°
Boiling Point: 160° dec.
Merck Reference: 14,554
MDL No.: MFCD00011429
Vzorec:
H₆N₂O₃S MW: 114,13 g/mol Bod varu: 160 °C (rozkl.) Bod tání: 131…135 °C Hustota: >1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011429 CAS číslo: 7773-06-0 EINECS: 231-871-7 Merck Index: 14,554 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...