Tisk…

Sulfamát amonný ≥98% ACS

Dodavatel: Thermo Scientific
Varování

Synonyms: Ammonium sulphamidate, Ammonium amidosulphonate

14534.36 14534.A3
14534.36EA 3300 CZK
14534.36 14534.A3
Sulfamát amonný ≥98% ACS
Ammonium sulphamate

Purity: 98% min CAS No.: 7773-06-0 Molecular Formula: H6N2O3S Formula Weight: 114.12 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Odour: Weak pH: 5.2 Melting Point: 131-135° Boiling Point: 160° dec. Solubility: Very soluble in water, liquid ammonia. Slightly soluble in ethanol, glycerol, glycol Merck Reference: 14,554 MDL No.: MFCD00011429


In electroplating solutions, making nitrous oxide gas, for manufacture of flameproofing textiles and paper products

Vzorec: H₆N₂O₃S
MW: 114,13 g/mol
Bod varu: 160 °C (rozkl.)
Bod tání: 131…135 °C
Hustota: >1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011429
CAS číslo: 7773-06-0
EINECS: 231-871-7
Merck Index: 14,554

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR