Tisk…

Kyselina sulfamová 99,99 Metals basis

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Aminosulphonic acid, Amidosulphuric acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid

44510.14 44510.22 44510.36
44510.14EA 1200 CZK
44510.14 44510.22 44510.36
Kyselina sulfamová 99,99 Metals basis
Kyselina sulfamová

Purity: 99.99% CAS No.: 5329-14-6 Molecular Formula: H2NSO3H Formula Weight: 97.09 Physical Form: Crystalline Density: 2.12 pH: 1.2 Melting Point: ca 205° dec. Merck Reference: 14,8921 UN No.: 2967 MDL No.: MFCD00011603

Vzorec: H₃NO₃S
MW: 97.09
Bod varu: 247
Bod tání: ∼ 205 °C (rozkl.)
Hustota: 2.12
Storage Temperature: Ambient
MDL Number: MFCD00011603
CAS číslo: 5329-14-6
EINECS: 226-218-8
UN: 2967
ADR: 8 III
Merck Index: 14,8921
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR