Kyselina sulfamová 99,3-100,3 Dried ACS
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Aminosulphonic acid, Amidosulphuric acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid
33233.22
33233.36
33233.22EA
1350
CZK
33233.22
33233.36
Kyselina sulfamová 99,3-100,3 Dried ACS
Kyselina sulfamová
Purity: 99.3-100.3%
CAS No.: 5329-14-6
Molecular Formula: H2NSO3H
Formula Weight: 97.09
Physical Form: Crystalline
Density: 2.12
pH: 1.2
Melting Point: ca 205° dec.
Merck Reference: 14,8921
UN No.: 2967
MDL No.: MFCD00011603
Vzorec:
H₃NO₃S MW: 97.09 Bod varu: 247 Bod tání: ∼ 205 °C (rozkl.) Hustota: 2.12 Storage Temperature: Ambient |
MDL Number:
MFCD00011603 CAS číslo: 5329-14-6 EINECS: 226-218-8 UN: 2967 ADR: 8 III Merck Index: 14,8921 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...