(Diethylamino)sulphur trifluoride 95%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
DAST
A11992.06
A11992.14
A11992.22
AB115869.0100
A11992.06EA
2930
CZK
A11992.06
A11992.14
A11992.22
ABCRAB115869.0100
(Diethylamino)sulphur trifluoride 95%
(Diethylamino)sulphur trifluoride
Purity: 95%
CAS No.: 38078-09-0
Molecular Formula: C4H10F3NS
Formula Weight: 161.19
Storage Temperature: 2°C to 8°C
Flash Point: 23°(73°F)
Refractive Index: 1.4115
Boiling Point: 30-32°/3mm
Beilstein Reference: 1849066
UN No.: 2920
MDL No.: MFCD00000363
Vzorec:
C₄H₁₀F₃NS Bod varu: 30…32 °C (3 mmHg) Hustota: 1,23 Bod vzplanutí: 23 °C (73 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000363 CAS číslo: 38078-09-0 EINECS: 253-771-2 UN: 2920 ADR: 8,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...