Tisk…

(Diethylamino)sulphur trifluoride 95%

Dodavatel: Thermo Scientific
Nebezpečí

216110050 216110250 216111000
ACRO216110050EA 3500 CZK
ACRO216110050 ACRO216110250 ACRO216111000
(Diethylamino)sulphur trifluoride 95%
(Diethylamino)sulphur trifluoride
Vzorec: C₄H₁₀F₃NS
Bod varu: 30…32 °C (3 mmHg)
Hustota: 1,316 g/cm³
Bod vzplanutí: 23 °C
Storage Temperature: Freezer
MDL Number: MFCD00000363
CAS číslo: 38078-09-0
EINECS: 253-771-2
UN: 2920
ADR: 8,I

Objednat

Výsledky specifikačního testu

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4160 (20°C, 589 nm)
NMR (1H-NMR) ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR