Seleninyl dichloride ≥99% (metals basis)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
68108.AB
68108.AC
68108.ABEA
12430
CZK
68108.AB
68108.AC
Seleninyl dichloride ≥99% (metals basis)
Seleninyl dichloride
Purity: 99%
CAS No.: 7791-23-3
Molecular Formula: SeOCl2
Formula Weight: 165.87
Physical Form: Liquid
Density: 2.42
Refractive Index: 1.651
Sensitivity: Moisture Sensitive
Appearance: Yellow
Melting Point: 8.5°
Boiling Point: 176.4°
Solubility: Decomposes in water forming HCl and H2SeO3
Merck Reference: 14,8435
UN No.: 2879
MDL No.: MFCD00016327
Vzorec:
Cl₂OSe MW: 165,86 g/mol Bod varu: 176,4 °C Bod tání: 8,5 °C Hustota: 2,42 Storage Temperature: Ambient |
MDL Number:
MFCD00016327 CAS číslo: 7791-23-3 EINECS: 232-244-0 UN: 2879 ADR: 8,I Merck Index: 14,8435 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...