Chlorosulphonyl isocyanate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
142662-100G
142662-1KG
142662-25G
SIAL142662-100GEA
4120
CZK
SIAL142662-100G
SIAL142662-1KG
SIAL142662-25G
Chlorosulphonyl isocyanate, Sigma-Aldrich®
Chlorosulphonyl isocyanate
Vzorec:
CClNO₃S Bod varu: 107 °C (1013 hPa) Bod tání: –44 °C Hustota: 1,62 g/cm³ (20 °C) |
MDL Number:
MFCD00011608 CAS číslo: 1189-71-5 EINECS: 214-715-2 UN: 3264 ADR: 8,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...