Tisk…

Síran amonný, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

M-Clarity™ quality level = MQ200

09978-1KG 09978-2.5KG 09978-500G
SIAL09978-1KGEA 6470 CZK
SIAL09978-1KG SIAL09978-2.5KG SIAL09978-500G
Síran amonný, Sigma-Aldrich®
Síran amonný
Vzorec: (NH₄)₂SO₄
Bod tání: 336…339 °C
Hustota: 1,78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS číslo: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR