Tisk…

Dihydrogenfosforečnan amonný ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

A15283.0C A15283.0E A15283.36 A15283.0G
A15283.0CEA 11910 CZK
A15283.0C A15283.0E A15283.36 A15283.0G
Dihydrogenfosforečnan amonný ≥98%
Dihydrogenfosforečnan amonný

Purity: 98% CAS No.: 7722-76-1 Molecular Formula: NH4H2PO4 Formula Weight: 115.03 Odour: Odourless Melting Point: 198-204° MDL No.: MFCD00003396

Vzorec: NH₄H₂PO₄
Bod tání: 198…204 °C
Hustota: 1,803
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS číslo: 7722-76-1
EINECS: 231-764-5
Merck Index: 14,543

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR